3-Chloro-5-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

074136-250MG 074136-1G 074136-5G
102534-830EA 355.38 USD
102534-830 102534-832 102534-834
3-Chloro-5-nitrobenzaldehyde ≥95%
3-Chloro-5-nitrobenzaldehyde

Hazard: Irritant

Formula: C₇H₄ClNO₃
MDL Number: MFCD08236780
CAS Number: 22233-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR