4-Hydroxy-3-(trifluoromethyl)benzoic acid ≥95%
Supplier: Matrix Scientific
074133-1G
074133-5G
074133-10G
102534-812EA
53.85
USD
102534-812
102534-814
102534-816
4-Hydroxy-3-(trifluoromethyl)benzoic acid ≥95%
4-Hydroxy-3-(trifluoromethyl)benzoic acid
Hazard: Irritant
Formula:
C₈H₅F₃O₃ MW: 206.12 g/mol |
MDL Number:
MFCD01091008 CAS Number: 220239-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...