2-Chloro-4-methoxybenzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-p-anisic acid

074131-1G 074131-5G 074131-10G
102534-800EA 178.46 USD
102534-800 102534-802 102534-804
2-Chloro-4-methoxybenzoic acid ≥95%
2-Chloro-4-methoxybenzoic acid

Hazard: Irritant

Formula: C₈H₇ClO₃
MDL Number: MFCD00085943
CAS Number: 21971-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR