2-Cyano-4-methoxybenzaldehyde ≥95%

Supplier: Matrix Scientific

074130-1G 074130-5G 074130-10G
102534-794EA 427.69 USD
102534-794 102534-796 102534-798
2-Cyano-4-methoxybenzaldehyde ≥95%
2-Cyano-4-methoxybenzaldehyde

Hazard: Irritant

Formula: C₉H₇NO₂
MDL Number: MFCD08275456
CAS Number: 21962-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR