5-Fluoro-m-toluonitrile ≥95%

Supplier: Matrix Scientific

074127-1G 074127-5G 074127-10G
102534-776EA 95.38 USD
102534-776 102534-778 102534-780
5-Fluoro-m-toluonitrile ≥95%
5-Fluoro-m-toluonitrile

Hazard: Irritant

Formula: C₈H₆FN
MW: 135.14 g/mol
MDL Number: MFCD03094312
CAS Number: 216976-30-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR