5-Bromo-2-thiophenecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-thiophenecarbonitrile, 5-Bromothiophene-2-carbonitrile, 2-Cyano-5-bromothiophene, 2-Bromo-5-cyanothiophene

074126-1G 074126-5G
102534-772EA 52.31 USD
102534-772 102534-774
5-Bromo-2-thiophenecarbonitrile ≥95%
5-Bromo-2-thiophenecarbonitrile

Hazard: Irritant

Formula: C₅H₂BrNS
MW: 188.0500 g/mol
MDL Number: MFCD04974080
CAS Number: 2160-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR