1-Aminocyclobutanecarboxylic acid methyl ester ≥95%

Supplier: Matrix Scientific

074125-1G 074125-5G
102534-768EA 461.54 USD
102534-768 102534-770
1-Aminocyclobutanecarboxylic acid methyl ester ≥95%
1-Aminocyclobutanecarboxylic acid methyl ester

Hazard: Irritant

Formula: C₆H₁₁NO₂
MDL Number: MFCD01318265
CAS Number: 215597-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR