6-Chloroanthranilic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-6-chlorobenzoic acid

074124-100G 074124-25G
102534-766EA 83.08 USD
102534-766 102663-978
6-Chloroanthranilic acid ≥95%
6-Chloroanthranilic acid

Hazard: Irritant

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Melting Pt: 149-152°C
MDL Number: MFCD00051530
CAS Number: 2148-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR