2-Chloro-4-methylbenzonitrile ≥95%

Supplier: Matrix Scientific

074121-1G 074121-5G
102534-750EA 92.31 USD
102534-750 102534-752
2-Chloro-4-methylbenzonitrile ≥95%
2-Chloro-4-methylbenzonitrile

Hazard: Irritant

Formula: C₈H₆CIN
MW: 151.5900 g/mol
MDL Number: MFCD03094672
CAS Number: 21423-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR