2-Chloro-3-fluoroaniline ≥95%

Supplier: Matrix Scientific

074120-1G 074120-5G 074120-10G
102534-744EA 72.31 USD
102534-744 102534-746 102534-748
2-Chloro-3-fluoroaniline ≥95%
2-Chloro-3-fluoroaniline

Hazard: Irritant

Formula: C₆H₅ClFN
MW: 145.56 g/mol
MDL Number: MFCD03407961
CAS Number: 21397-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR