4-Aminoimidazole-5-carboxamide ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-1H-imidazole-5-carboxamide, 5-Aminoimidazole-4-carboxamide, 5-Amino-4-imidazolecarboxamide, 5-Amino-1H-imidazole-4-carboxamide, 4-Aminoimidazole-5-carboxamide, 4-aminoimidazole-5-carboxamide, 5-Amino-3H-imidazole-4-Carboxamide

074118-1G 074118-5G 074118-10G
102534-732EA 15.38 USD
102534-732 102534-734 102534-736
4-Aminoimidazole-5-carboxamide ≥95%
4-Aminoimidazole-5-carboxamide

Hazard: Irritant

Formula: C₄H₆N₄O
MW: 126.12 g/mol
MDL Number: MFCD02181040
CAS Number: 360-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR