5,6-Dimethoxy-1-indanone ≥95%
Supplier: Matrix Scientific
074117-10G
074117-100G
102534-728EA
53.85
USD
102534-728
102534-730
5,6-Dimethoxy-1-indanone ≥95%
5,6-Dimethoxy-1-indanone
Hazard: Irritant
Formula:
C₁₁H₁₂O₃ |
MDL Number:
MFCD00003790 CAS Number: 2107-69-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...