6-Methyl-1H-indole-3-carboxylic acid ≥95%
Supplier: Matrix Scientific
074116-1G
074116-5G
102534-724EA
364.62
USD
102534-724
102534-726
6-Methyl-1H-indole-3-carboxylic acid ≥95%
6-Methyl-1H-indole-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₀H₉NO₂ MW: 175.1840 g/mol |
MDL Number:
MFCD06203633 CAS Number: 209920-43-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...