1-Benzyloxy-2-methyl-3-nitrobenzene ≥95%

Supplier: Matrix Scientific

074115-1G 074115-5G 074115-10G
102534-718EA 90.77 USD
102534-718 102534-720 102534-722
1-Benzyloxy-2-methyl-3-nitrobenzene ≥95%
1-Benzyloxy-2-methyl-3-nitrobenzene

Hazard: Irritant

Formula: C₁₄H₁₃NO₃
MW: 243.26 g/mol
MDL Number: MFCD00100652
CAS Number: 20876-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR