1-Methyl-2-piperidinemethanol ≥95%

Supplier: Matrix Scientific

074114-10G 074114-100G
102534-714EA 92.31 USD
102534-714 102534-716
1-Methyl-2-piperidinemethanol ≥95%
1-Methyl-2-piperidinemethanol

Hazard: Irritant

Formula: C₇H₁₅NO
MW: 129.2 g/mol
MDL Number: MFCD00006494
CAS Number: 20845-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR