(3-Chloro-5-fluorophenyl)acetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(3-Chloro-5-fluorophenyl)acetic acid

074110-1G 074110-5G 074110-10G
102534-694EA 178.46 USD
102534-694 102534-696 102534-698
(3-Chloro-5-fluorophenyl)acetic acid ≥95%
(3-Chloro-5-fluorophenyl)acetic acid

Hazard: Irritant

Formula: (C₆H₃ClF)CH₂COOH
MDL Number: MFCD03788475
CAS Number: 202001-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR