3-(Dimethylphosphono)-N-methylolpropionamide ≥95%

Supplier: Matrix Scientific

074108-1G 074108-5G 074108-10G
102534-682EA 107.69 USD
102534-682 102534-684 102534-686
3-(Dimethylphosphono)-N-methylolpropionamide ≥95%
3-(Dimethylphosphono)-N-methylolpropionamide

Hazard: Irritant

Formula: C₆H₁₄NO₅P
MDL Number: MFCD00468093
CAS Number: 20120-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR