(1S,2R)-cis-2-Aminocyclohexanol hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (1S,2R)-2-Aminocyclohexanol hydrochloride

074106-1G 074106-5G 074106-10G
102534-672EA 533.85 USD
102534-672 102534-674 102534-676
(1S,2R)-cis-2-Aminocyclohexanol hydrochloride ≥95%
(1S,2R)-cis-2-Aminocyclohexanol hydrochloride

Hazard: Irritant

Formula: C₆H₁₄ClNO
MDL Number: MFCD11618003
CAS Number: 200352-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR