2-Methyl-1-(naphthalen-2-yl)propan-2-amine ≥95%

Supplier: Matrix Scientific

074104-250MG 074104-1G 074104-5G
102534-660EA 640 USD
102534-660 102534-662 102534-664
2-Methyl-1-(naphthalen-2-yl)propan-2-amine ≥95%
2-Methyl-1-(naphthalen-2-yl)propan-2-amine

Hazard: Irritant

Formula: C₁₄H₁₇N
MW: 199.3 g/mol
MDL Number: MFCD11519113
CAS Number: 198226-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR