1-N-Cbz-pyrrolidine-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

074096-1G 074096-5G
102534-616EA 284.62 USD
102534-616 102534-618
1-N-Cbz-pyrrolidine-3-carboxylic acid ≥95%
1-N-Cbz-pyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₄
MDL Number: MFCD02179036
CAS Number: 188527-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR