2-Hydroxy-4-methylbenzonitrile ≥95%

Supplier: Matrix Scientific

074093-1G 074093-5G
102534-600EA 710.77 USD
102534-600 102534-602
2-Hydroxy-4-methylbenzonitrile ≥95%
2-Hydroxy-4-methylbenzonitrile

Hazard: Irritant

Formula: C₈H₇NO
MW: 133.1500 g/mol
MDL Number: MFCD02261941
CAS Number: 18495-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR