7-Chloro-4-chromanone ≥95%

Supplier: Matrix Scientific

074091-1G 074091-5G
102534-590EA 201.54 USD
102534-590 102534-592
7-Chloro-4-chromanone ≥95%
7-Chloro-4-chromanone

Hazard: Irritant

Formula: C₉H₇ClO₂
MW: 182.6000 g/mol
MDL Number: MFCD09744054
CAS Number: 18385-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR