3,5-Dichloropyridazine ≥95%

Supplier: Matrix Scientific

074090-1G 074090-5G
102534-586EA 72.31 USD
102534-586 102534-588
3,5-Dichloropyridazine ≥95%
3,5-Dichloropyridazine

Hazard: Irritant

Formula: C₄H₂Cl₂N₂
MW: 148.9781 g/mol
MDL Number: MFCD10698048
CAS Number: 1837-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR