2-Fluoro-6-nitroaniline ≥95%

Supplier: Matrix Scientific

074086-1G 074086-5G
102534-566EA 92.31 USD
102534-566 102534-568
2-Fluoro-6-nitroaniline ≥95%
2-Fluoro-6-nitroaniline

Hazard: Irritant

Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
MDL Number: MFCD04112583
CAS Number: 17809-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR