4-Cyano-3-fluorobenzoic acid ≥95%

Supplier: Matrix Scientific

074084-1G 074084-5G
102534-560EA 52.31 USD
102534-560 102534-562
4-Cyano-3-fluorobenzoic acid ≥95%
4-Cyano-3-fluorobenzoic acid

Hazard: Irritant

Formula: C₈H₄FNO₂
MW: 165.1213 g/mol
Melting Pt: 160…163 °C
MDL Number: MFCD03094324
CAS Number: 176508-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR