6-(Methylsulfonyl)-1,3-benzothiazol-2-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-6-(methylsulphonyl)benzothiazole

074083-1G 074083-5G 074083-10G
102534-554EA 46.15 USD
102534-554 102534-556 102534-558
6-(Methylsulfonyl)-1,3-benzothiazol-2-amine ≥95%
6-(Methylsulfonyl)-1,3-benzothiazol-2-amine

Hazard: Irritant

Formula: C8H8N2O2S2
CAS Number: 17557-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR