3-Cyclopropyl-1H-pyrazol-5-amine ≥95%

Supplier: Matrix Scientific

Synonyms: (5-Amino-1H-pyrazol-3-yl)cyclopropane, 5-Amino-3-cyclopropyl-1H-pyrazole, 3-Cyclopropyl-1H-pyrazol-5-amine

074082-1G 074082-5G
102534-550EA 178.46 USD
102534-550 102534-552
3-Cyclopropyl-1H-pyrazol-5-amine ≥95%
3-Cyclopropyl-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C₆H₉N₃
MW: 123.159 g/mol
MDL Number: MFCD08461681
CAS Number: 175137-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR