4-Bromo-3-(trifluoromethyl)benzonitrile ≥95%
Supplier: Matrix Scientific
074079-1G
074079-5G
102534-532EA
15.38
USD
102534-532
102534-534
4-Bromo-3-(trifluoromethyl)benzonitrile ≥95%
4-Bromo-3-(trifluoromethyl)benzonitrile
Hazard: Irritant
Formula:
C₈H₃BrF₃N |
MDL Number:
MFCD03095347 CAS Number: 1735-53-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...