5-Bromo-1,3-dichloro-2-fluorobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 3,5-Dichloro-4-fluorobromobenzene

074078-25G 074078-100G
102551-866EA 106.15 USD
102551-866 102551-868
5-Bromo-1,3-dichloro-2-fluorobenzene ≥95%
5-Bromo-1,3-dichloro-2-fluorobenzene

Hazard: Irritant/Light Sensitive

Formula: C₆H₂BrCl₂F
MW: 243.8900 g/mol
Boiling Pt: 234°C
Density: 1.824
Flash Pt: 95°C
MDL Number: MFCD09996901
CAS Number: 17318-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR