2-Amino-6-iodotoluene ≥95%

Supplier: Matrix Scientific

074077-1G 074077-5G 074077-10G
102534-520EA 355.38 USD
102534-520 102534-522 102534-524
2-Amino-6-iodotoluene ≥95%
2-Amino-6-iodotoluene

Hazard: Irritant

Formula: C₇H₈IN
MDL Number: MFCD08062502
CAS Number: 172681-47-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR