3-Methyl-4-isoxazolecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Methylisoxazole-4-carboxylic acid
074076-1G
074076-5G
102534-516EA
146.15
USD
102534-516
102534-518
3-Methyl-4-isoxazolecarboxylic acid ≥95%
3-Methyl-4-isoxazolecarboxylic acid
Hazard: Irritant
Formula:
C₅H₅NO₃ MW: 127.10 g/mol Melting Pt: 185…187 °C |
MDL Number:
MFCD01318161 CAS Number: 17153-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...