tert-Butyl-2-(hydroxymethyl)pyrrolidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Boc-DL-Prolinol, tert-Butyl 2-(hydroxymethyl)pyrrolidine-1-carboxylate

074075-1G 074075-5G 074075-10G
102534-510EA 346.15 USD
102534-510 102534-512 102534-514
tert-Butyl-2-(hydroxymethyl)pyrrolidine-1-carboxylate ≥95%
tert-Butyl-2-(hydroxymethyl)pyrrolidine-1-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₉NO₃
MW: 201.3 g/mol
MDL Number: MFCD01456556
CAS Number: 170491-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR