3-Benzyloxybenzyl bromide ≥95%
Supplier: Matrix Scientific
Synonyms:
3-(Benzyloxy)benzyl bromide, 1-(Benzyloxy)-3-(bromomethyl)benzene
074074-1G
074074-5G
102534-504EA
196.92
USD
102534-504
102534-506
3-Benzyloxybenzyl bromide ≥95%
3-Benzyloxybenzyl bromide
Hazard: Irritant
Formula:
C₁₄H₁₃BrO MW: 277.1600 g/mol |
MDL Number:
MFCD01318307 CAS Number: 1700-31-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...