tert-Butyl-4-(4-aminophenyl)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(tert-Butoxycarbonyl)-4-(4-aminophenyl)piperidine, 1-Boc-4-(4-aminophenyl)piperidine, tert-Butyl 4-(4-aminophenyl)piperidine-1-carboxylate, 4-(4-Aminophenyl)piperidine-1-carboxylic acid tert-Butyl ester

074073-1G 074073-5G
102534-500EA 186.15 USD
102534-500 102534-502
tert-Butyl-4-(4-aminophenyl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(4-aminophenyl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₆H₂₄N₂O₂
MW: 276.3740 g/mol
MDL Number: MFCD05861405
CAS Number: 170011-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR