4-(2-Aminoethyl)benzonitrile hydrochloride ≥95%

Supplier: Matrix Scientific

074071-1G 074071-5G 074071-10G
102534-486EA 184.62 USD
102534-486 102534-488 102534-490
4-(2-Aminoethyl)benzonitrile hydrochloride ≥95%
4-(2-Aminoethyl)benzonitrile hydrochloride

Hazard: Irritant

Formula: C₉H₁₁ClN₂
CAS Number: 167762-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR