5-Nitro-2-thiophenecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Nitrothiophene-2-carbonitrile

074070-1G 074070-5G 074070-10G
102534-480EA 46.15 USD
102534-480 102534-482 102534-484
5-Nitro-2-thiophenecarbonitrile ≥95%
5-Nitro-2-thiophenecarbonitrile

Hazard: Irritant

Formula: C₅H₂N₂O₂S
MW: 154.15 g/mol
MDL Number: MFCD00052401
CAS Number: 16689-02-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR