1-Bromo-2-chloro-5-nitrobenzene ≥95%
Supplier: Matrix Scientific
074069-10G
074069-100G
102534-476EA
107.69
USD
102534-476
102534-478
1-Bromo-2-chloro-5-nitrobenzene ≥95%
1-Bromo-2-chloro-5-nitrobenzene
Hazard: Irritant
Formula:
C₆H₃BrClNO₂ |
MDL Number:
MFCD00100437 CAS Number: 16588-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...