6-Nitro-o-anisidine ≥95%
Supplier: Matrix Scientific
Synonyms:
6-Nitro-o-anisidine, 2-Amino-3-nitroanisole, 2-Methoxy-6-nitrophenylamine, 2-Methoxy-6-nitroaniline
074068-1G
074068-5G
074068-10G
102534-470EA
73.85
USD
102534-470
102534-472
102534-474
6-Nitro-o-anisidine ≥95%
6-Nitro-o-anisidine
Hazard: Irritant
Formula:
C₇H₈N₂O₃ MW: 168.15 g/mol |
MDL Number:
MFCD01930197 CAS Number: 16554-45-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...