2-Cyano-4-methylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methylpyridine-2-carbonitrile, 4-Methylpicolinonitrile, 2-Cyano-γ-picoline, 2-Cyano-4-picoline, 4-Methyl-2-pyridinecarbonitrile

074065-1G 074065-5G 074065-10G
102534-454EA 32.31 USD
102534-454 102534-456 102534-458
2-Cyano-4-methylpyridine ≥95%
2-Cyano-4-methylpyridine

Hazard: Irritant

Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 145…148 °C (38 mmHg)
Melting Pt: 85…88 °C
MDL Number: MFCD00128868
CAS Number: 1620-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR