4-Bromo-3-(trifluoromethyl)benzoic acid ≥95%

Supplier: Matrix Scientific

074063-5G 074063-25G
102534-444EA 67.69 USD
102534-444 102663-974
4-Bromo-3-(trifluoromethyl)benzoic acid ≥95%
4-Bromo-3-(trifluoromethyl)benzoic acid

Hazard: Irritant

Formula: C₈H₄BrF₃O₂
MW: 269.02 g/mol
MDL Number: MFCD08459288
CAS Number: 161622-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR