DL-Phenylalaninol ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Amino-3-phenyl-1-propanol, DL-2-Amino-3-phenyl-1-propanol, 2-Amino-3-phenylpropan-1-ol
074062-1G
074062-5G
074062-10G
102534-436EA
46.15
USD
102534-436
102534-438
102534-440
DL-Phenylalaninol ≥95%
DL-Phenylalaninol
Hazard: Irritant
Formula:
C₉H₁₃NO MW: 151.21 g/mol |
MDL Number:
MFCD00066689 CAS Number: 16088-07-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...