3-(Chloromethyl)-5-methylpyridine hydrochloride ≥95%

Supplier: Matrix Scientific

074056-250MG 074056-1G 074056-5G
102534-402EA 364.62 USD
102534-402 102534-404 102534-406
3-(Chloromethyl)-5-methylpyridine hydrochloride ≥95%
3-(Chloromethyl)-5-methylpyridine hydrochloride

Hazard: Irritant

Formula: C7H9Cl2N
MW: 178.0600 g/mol
MDL Number: MFCD11656303
CAS Number: 1007089-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR