4-Bromo-2-nitro-6-(trifluoromethyl)aniline ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-bromo-3-nitrobenzotrifluoride

074054-1G 074054-5G 074054-10G
102534-390EA 533.85 USD
102534-390 102534-392 102534-394
4-Bromo-2-nitro-6-(trifluoromethyl)aniline ≥95%
4-Bromo-2-nitro-6-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₇H₄BrF₃N₂O₂
MW: 285.02 g/mol
MDL Number: MFCD09835318
CAS Number: 157026-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR