5-Bromo-o-toluonitrile ≥95%

Supplier: Matrix Scientific

074053-5G 074053-25G 074053-100G
102534-386EA 36.92 USD
102534-386 102663-970 102663-972
5-Bromo-o-toluonitrile ≥95%
5-Bromo-o-toluonitrile

Hazard: Irritant

Formula: C₈H₆BrN
MW: 196.05 g/mol
MDL Number: MFCD08061920
CAS Number: 156001-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR