Methyl-2-(4-bromophenyl)-2,2-dimethylacetate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-(4-bromophenyl)-2,2-dimethylacetate

074052-1G 074052-5G 074052-10G
102534-378EA 66.15 USD
102534-378 102534-380 102534-382
Methyl-2-(4-bromophenyl)-2,2-dimethylacetate ≥95%
Methyl-2-(4-bromophenyl)-2,2-dimethylacetate

Hazard: Irritant

Formula: C₁₁H₁₃BrO₂
MW: 257.1237 g/mol
MDL Number: MFCD02683357
CAS Number: 154825-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR