2,3-Dichloro-4-fluorobenzoic acid ≥95%

Supplier: Matrix Scientific

074050-250MG 074050-1G
102534-368EA 684.62 USD
102534-368 102534-370
2,3-Dichloro-4-fluorobenzoic acid ≥95%
2,3-Dichloro-4-fluorobenzoic acid

Hazard: Irritant

Formula: C₇H₃Cl₂FO₂
MW: 209.0000 g/mol
MDL Number: MFCD09835200
CAS Number: 154257-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR