4'-Ethylbiphenyl-4-ylboronic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
(4'-Ethyl-[1,1'-biphenyl]-4-yl)boronic acid
074048-1G
074048-5G
074048-10G
102534-356EA
213.85
USD
102534-356
102534-358
102534-360
4'-Ethylbiphenyl-4-ylboronic acid ≥95%
4'-Ethylbiphenyl-4-ylboronic acid
Hazard: Irritant
Formula:
C₁₄H₁₅BO₂ |
MDL Number:
MFCD04038068 CAS Number: 153035-62-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...