2-Fluoro-3-iodo-5-methylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-3-iodo-5-picoline

074047-1G 074047-5G
102534-352EA 533.85 USD
102534-352 102534-354
2-Fluoro-3-iodo-5-methylpyridine ≥95%
2-Fluoro-3-iodo-5-methylpyridine

Hazard: Irritant

Formula: C₆H₅FIN
MW: 237.02 g/mol
MDL Number: MFCD03095303
CAS Number: 153034-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR