4-Methylpyrimidin-2-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methylpyrimidin-2(1H)-one

074045-1G 074045-5G 074045-10G
102534-340EA 355.38 USD
102534-340 102534-342 102534-344
4-Methylpyrimidin-2-ol ≥95%
4-Methylpyrimidin-2-ol

Hazard: Irritant

Formula: C₅H₆N₂O
MW: 110.11 g/mol
MDL Number: MFCD09991726
CAS Number: 15231-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR