2-Fluoro-6-methoxybenzylamine ≥95%

Supplier: Matrix Scientific

Synonyms: (2-Fluoro-6-methoxyphenyl)methanamine

074039-1G 074039-5G
102534-310EA 246.15 USD
102534-310 102534-312
2-Fluoro-6-methoxybenzylamine ≥95%
2-Fluoro-6-methoxybenzylamine

Hazard: Irritant

Formula: C₈H₁₀FNO
MW: 155.1695 g/mol
MDL Number: MFCD00042292
CAS Number: 150517-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR